ContactUsLinkComponent
Guzzler 44 Hose for model 07090-10 (Tube length: 192)
Guzzler 44 Hose for model 07090-10 (Tube length: 192)
# de catalogue MFLX06309-10
Fournisseur:  BOSWORTH CO
Guzzler 44 Hose for model 07090-10 (Tube length: 192)
Guzzler 44 Hose for model 07090-10 (Tube length: 192)
# de catalogue MFLX06309-10
Fournisseur:  BOSWORTH CO
  • Warranty:
  • 1 year
  • 1 year
  • En cours de visualisation
Spécifications
A propos de cet article
Optimize performance of your Guzzler® Diaphragm Hand Pump.
  • Suitable for use with Guzzler® Portable Diaphragm Hand Pumps
These hoses are noncollapsing, wire-reinforced, and constructed of durable vinyl.
Model 06309-10 hose is compatible with Guzzler pumps 07090-09 and -10.
Model 06309-11 hose is compatible with Guzzler pump 07090-11.
Model 06309-12 hose is compatible with Guzzler pump 07090-12.
  • Description:
    Hose for model 07090-10 (Tube length: 192)
  • Warranty:
    1 year
  • Cat. no.:
    MFLX06309-10
Customers Who Bought This Also Bought