ContactUsLinkComponent
GRINDING BALL SS 1/8IN (3 MM) PK1000
Cole-Parmer® Grinding Balls
# de catalogue 76533-062
Fournisseur:  SPEX
GRINDING BALL SS 1/8IN (3 MM) PK1000
Cole-Parmer® Grinding Balls
# de catalogue 76533-062
Fournisseur:  SPEX
Spécifications
  • Description:
    Grinding balls , pack of 1000
  • Material:
    Stainless steel
  • Size:
    3 mm (¹/₈")
  • Cat. no.:
    76533-062
  • For Use With:
    Spex® SamplePrep homogenizers, grinders, and mills
Spécifications
A propos de cet article
Crush, grind, and mill various materials.
  • Use grinding balls to grind samples in Cole-Parmer® homogenizers and grinders
Replace vial set grinding balls to ensure optimal grinding potential.
Customers Who Bought This Also Bought