Watch Glasses, Economy, United Scientific Supplies
Catalog # 76532-272
Supplier: United Scientific Supplies
Specifications
- Ø:300 mm
- Description:Watch glasses, economy
- Cat. no.:76532-272
Specifications
About this item
Useful clear watch glasses for evaporation.
- Flint glass
- Flame polished edges
Made from clear flint glass (1.8 to 2.0mm thick), fine ground and polished edges.
Frequently Bought Together