Watch Glasses, Economy, United Scientific Supplies
# de catalogue 76532-260
Fournisseur: United Scientific Supplies
Spécifications
- Ø:40 mm
- Description:Watch glasses, economy
- Cat. no.:76532-260
Spécifications
A propos de cet article
Useful clear watch glasses for evaporation.
- Flint glass
- Flame polished edges
Made from clear flint glass (1.8 to 2.0mm thick), fine ground and polished edges.
Customers Who Bought This Also Bought