Pipettes, Transfer, Volumetric, Class A, Individually Certified, United Scientific Supplies
# de catalogue 76532-140
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:25 ml
- Description:Blue, 0.030 ± ml tolerance
- Cat. no.:76532-140
Spécifications
A propos de cet article
Volumetric pipettes with color-coding.
- Printed with durable amber markings
- Color-coded in blue for easy identification of capacity
Made from borosilicate glass in compliance with ASTM E969, Class A standards. Individually serialized and certified; calibrated to deliver.
Customers Who Bought This Also Bought