Order Entry
ContactUsLinkComponent
TUBING FLINT GLASS 1LB 24X5MM
Glass Tubing, Flint, United Scientific Supplies
# de catalogue 76532-026
TUBING FLINT GLASS 1LB 24X5MM
Glass Tubing, Flint, United Scientific Supplies
# de catalogue 76532-026

Spécifications

  • Description:
    Glass Tubing, Flint, 24" × 5 mm
  • Cat. no.:
    76532-026

Spécifications

A propos de cet article

Tubing made from fine quality flint glass for easy bending, blowing and sealing of soft glass apparatus.

  • Borosilicate glass tubing

Tubing is 24" long and available in various diameters. Sold per pound.

Customers Who Bought This Also Bought