Order Entry
ContactUsLinkComponent
FLASK VOLUMETRIC CLASS A 1000ML
Volumetric Flasks, Class A, with Glass Stopper, Batch Certified, United Scientific Supplies
# de catalogue 76531-866
Volumetric Flasks, Class A, with Glass Stopper, Batch Certified, United Scientific Supplies
# de catalogue 76531-866

Spécifications

  • Capacity:
    1000 ml
  • Cat. no.:
    76531-866
  • Stopper No.:
    22
  • Tolerance:
    ±0.30 ml

Spécifications

A propos de cet article

Durable Class A Volumetric flasks.

  • Sharp graduation line and large white block lettering

Durable, machine-blown body. Heavy beaded tubing neck.

Certifications: Complies with ASTM E288 Class A specifications; batch certified.
Ordering information: A ground glass stopper is included.
Customers Who Bought This Also Bought