Order Entry
ContactUsLinkComponent
FLASK VOLUMETRIC CLASS A 50ML
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, United Scientific Supplies
# de catalogue 76531-852
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, United Scientific Supplies
# de catalogue 76531-852

Spécifications

  • Capacity:
    50 ml
  • Cat. no.:
    76531-852
  • Stopper No.:
    9
  • Tolerance:
    ±0.05 ml

Spécifications

A propos de cet article

Durable, machine-blown body with heavy beaded tubing neck.

  • Sharp graduation line and large white block lettering

A ground glass stopper is included.

Certifications: Complies with ASTM E288 Class A specifications; individually serialized and certified.
Customers Who Bought This Also Bought