ContactUsLinkComponent
CYLINDER GLASS GRADUATED 1000ML
Graduated Cylinders, Glass, Class A, Batch Certified, with Stopper, United Scientific Supplies
# de catalogue 76531-574
Graduated Cylinders, Glass, Class A, Batch Certified, with Stopper, United Scientific Supplies
# de catalogue 76531-574
Spécifications
  • Capacity:
    1000 ml
  • Graduation Interval:
    10 ml
  • Cat. no.:
    76531-574
  • Tolerance:
    ±3.0 ml
Spécifications
A propos de cet article
Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing and strong, stable, hexagonal bases. All cylinders come with bumper guards.
  • With hollow glass stopper
  • Double metric scale, calibrated to contain
Cylinders comply with ASTM E-1272, Class A standards, batch certified.
Customers Who Bought This Also Bought