ContactUsLinkComponent
BURETTE GLASS PTFE STOPCOCK 10ML
Burettes, Class B, with PTFE Detachable Stopcocks, United Scientific Supplies
# de catalogue 76531-450
BURETTE GLASS PTFE STOPCOCK 10ML
Burettes, Class B, with PTFE Detachable Stopcocks, United Scientific Supplies
# de catalogue 76531-450
Spécifications
  • Capacity:
    10 ml
  • Graduation Interval:
    0.05 ml
  • Description:
    Burette, class B, PTFE detachable stopcock
  • Cat. no.:
    76531-450
Spécifications
A propos de cet article
Made from high-quality accurate bore tubing for a high degree of accuracy, reliability and durability.
  • Class B burette
  • Graduated at intervals
  • Available in multiple sizes
Detachable PTFE stopcock and tip facilitate easy cleaning and convenient replacement of broken tips.
Customers Who Bought This Also Bought