Order Entry
ContactUsLinkComponent
BOTTLE MEDIA GLASS AMBER 1000ML PK10
Media/Storage Bottles, Amber, United Scientific Supplies
Catalog # 76531-432
BOTTLE MEDIA GLASS AMBER 1000ML PK10
Media/Storage Bottles, Amber, United Scientific Supplies
Catalog # 76531-432

Specifications

  • Graduation range:
    100 - 900
  • Capacity:
    1000 ml
  • Description:
    Media/storage bottles, amber
  • Cat. no.:
    76531-432
  • O.D.×H:
    203×101 mm

Specifications

About this item

Pack of 10 borosilicate glass media / storage bottles.

  • Amber color designed to protect light-sensitive materials during transport and storage
  • Feature printed white graduations (from 40 - 80 ml) and durable marking spots
  • Maximum use temperature is 450 °C (842 °F)
  • Autoclavable

Included GL 45 cap is linerless, both the caps and drip-free rings are made from polypropylene and autoclavable to 140 °C.

Customers Who Bought This Also Bought