ContactUsLinkComponent
BOTTLE DROPPING SQUARE GLASS 15ML PK12
Bottles With Dropper, Square, Flint Glass, United Scientific Supplies
# de catalogue 76531-322
BOTTLE DROPPING SQUARE GLASS 15ML PK12
Bottles With Dropper, Square, Flint Glass, United Scientific Supplies
# de catalogue 76531-322
Spécifications
  • Capacity:
    0.5 oz.
  • Color:
    Clear
  • Description:
    Bottles with dropper, square, flint glass
  • Material:
    Flint glass
  • Cat. no.:
    76531-322
Spécifications
A propos de cet article
Pack of 12 square glass dropping bottles.
  • Non-porous glass is easy to clean and autoclavable
These square flint glass bottles include screw cap with dropper assembly are designed for general use. The glass is non-porous, which allows for easy cleaning or autoclaving.
Ordering information: Screw cap with dropper assembly included.
Customers Who Bought This Also Bought