Ward's® Live Waxworms (Galleria mellonella)
# de catalogue 470336-812
Fournisseur: DMF BAIT COMPANY
Spécifications
- Description:Pack of 250
- Cat. no.:470336-812
Spécifications
A propos de cet article
Wax worms are a hardy larvae that are easy to handle.
- Chemical-free, safe for feeding to pets
- Healthy for many reptiles and fish
- These creamy white worms have soft, grublike bodies and are dry to the touch
- Refrigeration optional
Wax worms do not need moisture, and you do not need to feed them as in their larva state they live off their own body fat. You can keep them at room temperature, or around 45 to 65 °F for maximum shelf life.
Delivery: These products contain living materials and will ship via overnight or 2 nd day delivery. Please specify a delivery date during checkout.
Packaging: Packed in cups with wood chips.
Customers Who Bought This Also Bought