ContactUsLinkComponent
Brush Long Handle 16.54in Hard White, Easily clean small spaces with, 1.73in wide narrow-headed brush, bristles are angled to facilitate easy cleaning corners, grip is desinged for extra stability and, entire brush is color-coded to support HACCP principles.
Vikan® Long Handle, Narrow Head Cleaning Brushes, Remco
# de catalogue 76530-426
Fournisseur:  Remco Products
Brush Long Handle 16.54in Hard White, Easily clean small spaces with, 1.73in wide narrow-headed brush, bristles are angled to facilitate easy cleaning corners, grip is desinged for extra stability and, entire brush is color-coded to support HACCP principles.
Vikan® Long Handle, Narrow Head Cleaning Brushes, Remco
# de catalogue 76530-426
Fournisseur:  Remco Products
Spécifications
  • Description:
    Long handle, narrow head cleaning brush, white
  • Cat. no.:
    76530-426
Spécifications
A propos de cet article
Easily clean small spaces with this narrow-headed cleaning brush.
  • Polyester bristles, polypropylene block
The bristles are angled to allow easy cleaning of corners, the grip is designed for extra stability and the entire brush is color-coded to support HACCP principles.
Overall dimensions: 16.54L×1.81W×4.33"H
Customers Who Bought This Also Bought