Vikan® Single-Blade Ultra-Hygiene Squeegees, Remco
# de catalogue 76530-362
Fournisseur: Remco Products
Spécifications
- Color:Brown
- Cat. no.:76530-362
- Blade Width:16"
Spécifications
A propos de cet article
The one-piece, single-blade squeegee has an ultra-hygienic design that minimizes areas for water and food soils to collect. This seamless, one-piece construction is especially effective in areas requiring the highest level of hygiene.
- Rubber blade, polypropylene block
- Seamless design
- Euro (hygienic) thread type
- Available in three blade widths
The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces. Hygiene squeegees are a good foundation for any HACCP or food safety program.
Customers Who Bought This Also Bought