ContactUsLinkComponent
Squeegee Ultra Hygiene 16in Pp/Rb Brown, one-piece, single blade squeegee has an ultra-hygienic design that minimizes areas for water and food soils to collect, seamless, one-piece contstruction is especially effective in areas requiring, highest level of hygiene. Good foundation for any HACCP
Vikan® Single-Blade Ultra-Hygiene Squeegees, Remco
# de catalogue 76530-362
Fournisseur:  Remco Products
Squeegee Ultra Hygiene 16in Pp/Rb Brown, one-piece, single blade squeegee has an ultra-hygienic design that minimizes areas for water and food soils to collect, seamless, one-piece contstruction is especially effective in areas requiring, highest level of hygiene. Good foundation for any HACCP
Vikan® Single-Blade Ultra-Hygiene Squeegees, Remco
# de catalogue 76530-362
Fournisseur:  Remco Products
Spécifications
  • Color:
    Brown
  • Cat. no.:
    76530-362
  • Blade Width:
    16"
Spécifications
A propos de cet article
The one-piece, single-blade squeegee has an ultra-hygienic design that minimizes areas for water and food soils to collect. This seamless, one-piece construction is especially effective in areas requiring the highest level of hygiene.
  • Rubber blade, polypropylene block
  • Seamless design
  • Euro (hygienic) thread type
  • Available in three blade widths
The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces. Hygiene squeegees are a good foundation for any HACCP or food safety program.
Customers Who Bought This Also Bought