ContactUsLinkComponent
Brush With Long Handle 23.62in Md Blue, with angled Bristle Security Units is effective with cleaning conveyor belts, production lines, machinery, and food preperation surfaces in high risk areas.
Vikan® Ultra-Slim Wand Brushes, Remco
# de catalogue 76528-960
Fournisseur:  Remco Products
Brush With Long Handle 23.62in Md Blue, with angled Bristle Security Units is effective with cleaning conveyor belts, production lines, machinery, and food preperation surfaces in high risk areas.
Vikan® Ultra-Slim Wand Brushes, Remco
# de catalogue 76528-960
Fournisseur:  Remco Products
Spécifications
  • Description:
    Ultra-slim wand brush, blue
  • Cat. no.:
    76528-960
Spécifications
A propos de cet article
Clean where other brushes cannot reach with this ultra-slim (15 mm wide) brush designed with a scraping edge for tackling stubborn debris.
  • Color-coded to support HACCP principles
  • Regenerated polypropylene block, polyester bristles
  • Food contact compliant
This brush features an easy-grip slim handle and a specially designed head with extra-stiff first bristles that help clean all the way to the end of narrow spaces. The brush block is made using food-contact compliant polypropylene offcuts from other Vikan products.
Overall dimensions: 23.62L×0.59W×2.36"H
Customers Who Bought This Also Bought