ContactUsLinkComponent
Brush Utility Medium 10.2in Pp/Pbt Blue, small, light-weight brush has a broad head and an ergonomically angled handle, design raises, user's hand from, cleaning surface reducing, risk of injury. Suitable for cleaning small surface areas, tables, pots, cutting boards, etc.
Vikan® Small Utility Brush, Medium Bristles, Remco
# de catalogue 76528-788
Fournisseur:  Remco Products
Brush Utility Medium 10.2in Pp/Pbt Blue, small, light-weight brush has a broad head and an ergonomically angled handle, design raises, user's hand from, cleaning surface reducing, risk of injury. Suitable for cleaning small surface areas, tables, pots, cutting boards, etc.
Vikan® Small Utility Brush, Medium Bristles, Remco
# de catalogue 76528-788
Fournisseur:  Remco Products
Spécifications
  • Description:
    Small utility brush, blue
  • Cat. no.:
    76528-788
Spécifications
A propos de cet article
Small, lightweight brush features a broad head and an ergonomically angled handle.
  • Suitable for cleaning small surface areas, tables, pots, cutting boards
  • Polypropylene block, polyester bristles
Design raises the user's hand from the cleaning surface reducing the risk of injury.
Overall dimensions: 10.24L×1.57W×2.76"H
Customers Who Bought This Also Bought