Kit, Exiprep 96 Viral Dna/Rna, Pre-filled buffer cartridge system, Simultaneous extraction of nucleic acids from up to 96 clinical samples, instrument that automatically extracts nucleic acids using magnetic rods
# de catalogue 76522-160
Fournisseur: Bioneer
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.