Exiprep 96 Lite, Auto Nucleic acid Extraction System, that automatically extracts nucleic acids from up to 96 samples, Magnetic rod method for extracting nucleic acids within 30 minutes, can easily extract the nucleic acid, various samples such as blood and serum can be applied
# de catalogue 76522-158
Fournisseur: Bioneer
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.