Smart-CO2 sensor, with temp and humidity sensors for use with Smart-Vue Pro systems. CO2 range: 0.5% to 9.9%. Temp range: 0 deg C to +50 deg C, Relative humidity range: 0% to 99%. 5V external power supply. Calibrated at 5%CO2, 37 deg C and 80% RH.
# de catalogue 76520-802
Fournisseur: Thermo Fisher Scientific
Customers Who Bought This Also Bought