VWR Controller, constant-temperature system a1201, used for p/pv1-6
# de catalogue 76520-732
Fournisseur: VWR International
VWR Controller, constant-temperature system a1201, used for p/pv1-6
# de catalogue 76520-732
Fournisseur: VWR International
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
881,80 $ to 2 807,56 $
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
458,88 $ to 1 237,78 $
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Fournisseur: MilliporeSigma
1 352,49 $
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Fournisseur: Advanced Instruments
2 423,69 $
Sol-Care® Safety Winged Blood Collection Needles, Sol M
Fournisseur: SOL-MILLENNIUM MEDICAL
84,04 $ to 651,48 $