Diamond® SureFlow™ Bottle Top Dispensers, Globe Scientific
Catalog # 76520-482
Supplier: Globe Scientific
Specifications
- Volume:5.0 - 60 ml
- Cat. no.:76520-482
- Increments:1.0 ml
Specifications
About this item
Diamond® SureFlow™ Bottle Top Dispensers have been designed with several features to make the dispensing of valuable laboratory reagents easier than ever.
- Two-year warranty
- Fully autoclavable
- Single tool recalibration
- Locking feature to prevent spills
- 360° rotation to dispense in multiple vessels
- Lower pressure for larger volumes helps reduce splashing
- Telescopic fill tube for bottles ranging in height from 170 to 300 mm
With a choice of standard or recirculating models, bottle top dispensers include a unique piston design which allows cleaning and servicing without changing the calibration, and a springless valve which requires less maintenance, offering higher reliability and added chemical resistance. Dual graduations, volume lock, and four additional bottle thread adapters are all standard. Recirculating models help eliminate the loss of valuable reagents by priming without loss.
Customers Who Bought This Also Bought