Order Entry
ContactUsLinkComponent
Bottle Top Dispenser, Standard, 5.0mL to 50.0mL, 1.0mL increments
Diamond® SureFlow™ Bottle Top Dispensers, Globe Scientific
# de catalogue 76520-464
Fournisseur:  Globe Scientific
Bottle Top Dispenser, Standard, 5.0mL to 50.0mL, 1.0mL increments
Diamond® SureFlow™ Bottle Top Dispensers, Globe Scientific
# de catalogue 76520-464
Fournisseur:  Globe Scientific

Spécifications

  • Volume:
    5.0 - 50 ml
  • Cat. no.:
    76520-464
  • Increments:
    1.0 ml

Spécifications

A propos de cet article

Diamond® SureFlow™ Bottle Top Dispensers have been designed with several features to make the dispensing of valuable laboratory reagents easier than ever.

  • Two-year warranty
  • Fully autoclavable
  • Single tool recalibration
  • Locking feature to prevent spills
  • 360° rotation to dispense in multiple vessels
  • Lower pressure for larger volumes helps reduce splashing
  • Telescopic fill tube for bottles ranging in height from 170 to 300 mm

With a choice of standard or recirculating models, bottle top dispensers include a unique piston design which allows cleaning and servicing without changing the calibration, and a springless valve which requires less maintenance, offering higher reliability and added chemical resistance. Dual graduations, volume lock, and four additional bottle thread adapters are all standard. Recirculating models help eliminate the loss of valuable reagents by priming without loss.

Customers Who Bought This Also Bought