Mobile Clear Bin Storage Unit 16S, Double-Row holds 16 3in tubs, each capable of holding up to 20 lbs. The steel frame & sturdy guardrails with built-in locking system keeps the tough polypropylene bins securely in place during storage or transit, preventing them from unintentionally being removed
# de catalogue 76519-984
Fournisseur: Luxor
Customers Who Bought This Also Bought