VWR® Temperature Monitoring Device
# de catalogue 76520-150
Fournisseur: VWR International
Spécifications
- Description:Temperature monitoring device
- Weight:1.1 kg (2.5 lbs.)
- Exterior W×D×H:203×127×76 mm (8×5×3")
- Cat. no.:76520-150
Spécifications
A propos de cet article
This temperature monitoring device comes with a buffered probe in product simulated solution. It includes an °F/°C switchable readout, audible and visual alarms, and a 3 year certificate of calibration.
- Material: Assorted materials
Customers Who Bought This Also Bought