ContactUsLinkComponent
Magnetic stirrer bars, 6 mm long x 3 mm diameter, PTFE-coated. Suitable for use with spectrophotometer cells designed specifically to accommodate magnetic stir bars (such as p/n 6610015400)
Magnetic Stirring Bars for UV-Vis and UV-Vis-NIR, Agilent Technologies
# de catalogue CAAG6610018900
Magnetic stirrer bars, 6 mm long x 3 mm diameter, PTFE-coated. Suitable for use with spectrophotometer cells designed specifically to accommodate magnetic stir bars (such as p/n 6610015400)
Magnetic Stirring Bars for UV-Vis and UV-Vis-NIR, Agilent Technologies
# de catalogue CAAG6610018900
Spécifications
  • Coating:
    PTFE
  • Description:
    Magnetic stirrer bars, 6 mm long × 3 mm diameter, PTFE-coated. Suitable for use with spectrophotometer cells designed specifically to accommodate magnetic stir bars (includes p/n 6610015400)
  • Cat. no.:
    CAAG6610018900
Spécifications
A propos de cet article
Magnetic stir bars or star type stirrers provide uniform & thorough mixing of samples in cuvettes.
  • Magnetic stirrers are designed specifically for use with standard rectangular cells in cell holders that have magnetic stirring capability
  • Suitable for use with standard 10×10 mm cells or the specialized micro cell that includes a circular recess in the base for the stir bar (pn 6610015400)
  • Multi-faceted stirrer profile provides rapid vertical and horizontal mixing with a minimum of vortexing
  • Smooth surface ensures the stirrer bar stays close to the bottom of the vessel
  • Optimal coupling as the length of the stir bar is in close contact with magnetic drive mechanism
  • Alnico magnet encapsulated in PTFE coating for superior chemical resistance
  • Stir bars are autoclavable
Magnetic stir bars or star type stirrers provide uniform and thorough mixing of samples in cuvettes. The stir bars are recommended for the stirring module and Peltier thermostatted (1×1) cell holder. The star stirrer is recommended for Peltier thermostatted multicell holders for Cary UV-Vis systems.
Customers Who Bought This Also Bought