ASPIR-8™ Solution Reservoirs
# de catalogue 470336-444
Fournisseur: MTC BIO, INC.
Spécifications
- Capacity:5 ml
- Sterility:Non sterile
- Description:Reservoirs, bulk pack
- Cat. no.:470336-444
Spécifications
A propos de cet article
ASPIR-8™ is the only reservoir that is tailor-made for your 8-channel instrument.
- Specially designed for 8-channel pipettes
- 75 mm inner chamber
- Conserve valuable reagents
Our new ASPIR-8™ is the ideal reservoir for any lab utilizing 8 channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over its longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage.
Customers Who Bought This Also Bought