Gown, Chemotherapy, Level 2, Blue, Full front, long sleeves, waist ties, knitted cuffs, and a full back for complete coverage, Ultrasonically sealed seams for superior protection and high tensile strength, soft, multilayer nonwoven spunbond material with fluid-impervious PE coating, Size: 2Xl
# de catalogue 76515-536
Fournisseur: Tronex
Customers Who Bought This Also Bought