Azure B, Molecular Weight: 305.83, CAS Number: 531-55-5, Chemical Formula: C15H16ClN3S, Solubility: 0.1% (Distilled Water), Common Name: Azure I, Size: 25 G
# de catalogue 76509-148
Fournisseur: RPI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.