ContactUsLinkComponent
Lab Pack Poly Drum, 55 Gal, Metal Bolt Ring, 2x2in Bung Holes, Black, Dimensions, Exterior: 21in (53.3 cm) Top, 22.5in (57.2 cm) Bottom, 36.375in (92.4 cm) Height
# de catalogue 76502-718
Fournisseur:  Eagle Manufacturing
Lab Pack Poly Drum, 55 Gal, Metal Bolt Ring, 2x2in Bung Holes, Black, Dimensions, Exterior: 21in (53.3 cm) Top, 22.5in (57.2 cm) Bottom, 36.375in (92.4 cm) Height
Lab Pack Poly Drum, 55 Gal, Metal Bolt Ring, 2x2in Bung Holes, Black, Dimensions, Exterior: 21in (53.3 cm) Top, 22.5in (57.2 cm) Bottom, 36.375in (92.4 cm) Height
# de catalogue 76502-718
Fournisseur:  Eagle Manufacturing
Customers Who Bought This Also Bought