Lab Pack Open Head Poly Drum, 55 Gal, Metal Lever-Lock, 1x2in 1x3/4in Bung Holes, Blue, Dimensions, Exterior: 21in (53.3 cm) Top, 22.5in (57.2 cm) Bottom, 36.375in (92.4 cm) Height
# de catalogue 76502-704
Fournisseur: Eagle Manufacturing
Customers Who Bought This Also Bought