ContactUsLinkComponent
Lab Pack Open Head Poly Drum, 40 Gal, Metal Lever-Lock, Black, Dimensions, Exterior: 23.75in (60.3 cm) Top, 19.13in (48.6 cm) Bottom, 31.13in (79.1 cm) Height
# de catalogue 76502-688
Fournisseur:  Eagle Manufacturing
Lab Pack Open Head Poly Drum, 40 Gal, Metal Lever-Lock, Black, Dimensions, Exterior: 23.75in (60.3 cm) Top, 19.13in (48.6 cm) Bottom, 31.13in (79.1 cm) Height
Lab Pack Open Head Poly Drum, 40 Gal, Metal Lever-Lock, Black, Dimensions, Exterior: 23.75in (60.3 cm) Top, 19.13in (48.6 cm) Bottom, 31.13in (79.1 cm) Height
# de catalogue 76502-688
Fournisseur:  Eagle Manufacturing
Customers Who Bought This Also Bought