ContactUsLinkComponent
Lab Pack Open Head Poly Drum, 14 Gal, Metal Bolt Ring, 1x2in 1x3/4in Bung Holes, Black, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 26.5in (67.3 cm) Height
# de catalogue 76502-636
Fournisseur:  Eagle Manufacturing
Lab Pack Open Head Poly Drum, 14 Gal, Metal Bolt Ring, 1x2in 1x3/4in Bung Holes, Black, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 26.5in (67.3 cm) Height
Lab Pack Open Head Poly Drum, 14 Gal, Metal Bolt Ring, 1x2in 1x3/4in Bung Holes, Black, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 26.5in (67.3 cm) Height
# de catalogue 76502-636
Fournisseur:  Eagle Manufacturing
Customers Who Bought This Also Bought