VWR® Inserts for Wide Opening Vials
Catalog # 82028-452A
Supplier: VWR International
Specifications
- Cap included:No
- Capacity:0.40 ml
- Color:Clear
- Description:Insert, pulled point bottom, for wide opening vials
- Material:Glass
- Cat. no.:82028-452A
- Bottom Style:Conical bottom