VWR® Manual Crimping and Decapping Tools
# de catalogue 76047-570A
Fournisseur: VWR International
Spécifications
- Description:Manual decapper, steel, black
- Cat. no.:76047-570A
- Seal Size:20 mm
Spécifications
A propos de cet article
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
- Easy and convenient handling
- Durable, high quality construction
- Adjustable screw setting facilitates optimum crimping
Adjustable in crimping pressure limitation by screw in handle.
Customers Who Bought This Also Bought