ContactUsLinkComponent
VWR® Manual Crimping and Decapping Tools
  76047-562A
VWR® Manual Crimping and Decapping Tools
  76047-562A
  • Description:
    Manual crimper, steel, black
  • Cat. no.:
    76047-562A
  • Seal Size:
    20 mm
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
  • Easy and convenient handling
  • Durable, high quality construction
  • Adjustable screw setting facilitates optimum crimping
Adjustable in crimping pressure limitation by screw in handle.
Frequently Bought Together