VWR® Manual Crimping and Decapping Tools
76047-562A
- Description:Manual crimper, steel, black
- Cat. no.:76047-562A
- Seal Size:20 mm
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
- Easy and convenient handling
- Durable, high quality construction
- Adjustable screw setting facilitates optimum crimping
Adjustable in crimping pressure limitation by screw in handle.
Frequently Bought Together