ContactUsLinkComponent
VWR Manual Crimper, Seal size: 11 mm, provide secure vial closure, Easy and convenient handling, Durable, high quality construction
VWR® Manual Crimping and Decapping Tools
Catalog # 76047-558A
Supplier:  VWR International
VWR® Manual Crimping and Decapping Tools
Catalog # 76047-558A
Supplier:  VWR International
Specifications
  • Description:
    Manual crimper, steel, black
  • Cat. no.:
    76047-558A
  • Seal Size:
    11 mm
Specifications
About this item
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
  • Easy and convenient handling
  • Durable, high quality construction
  • Adjustable screw setting facilitates optimum crimping
Adjustable in crimping pressure limitation by screw in handle.
Customers Who Bought This Also Bought