VWR® Manual Crimping and Decapping Tools
Catalog # 76047-558A
Supplier: VWR International
Specifications
- Description:Manual crimper, steel, black
- Cat. no.:76047-558A
- Seal Size:11 mm
Specifications
About this item
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
- Easy and convenient handling
- Durable, high quality construction
- Adjustable screw setting facilitates optimum crimping
Adjustable in crimping pressure limitation by screw in handle.
Customers Who Bought This Also Bought