ContactUsLinkComponent
Lab Pack Open Head Poly Drum, 10 Gal, Metal Lever-Lock, Yellow, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 18.333in (46.6 cm) Height
# de catalogue 76502-608
Fournisseur:  Eagle Manufacturing
Lab Pack Open Head Poly Drum, 10 Gal, Metal Lever-Lock, Yellow, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 18.333in (46.6 cm) Height
Lab Pack Open Head Poly Drum, 10 Gal, Metal Lever-Lock, Yellow, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 18.333in (46.6 cm) Height
# de catalogue 76502-608
Fournisseur:  Eagle Manufacturing
Customers Who Bought This Also Bought