Lab Pack Open Head Poly Drum, 10 Gal, Metal Lever-Lock, Yellow, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 18.333in (46.6 cm) Height
# de catalogue 76502-608
Fournisseur: Eagle Manufacturing
Lab Pack Open Head Poly Drum, 10 Gal, Metal Lever-Lock, Yellow, Dimensions, Exterior: 15in (38.1 cm) Top, 12.75in (32.4 cm) Bottom, 18.333in (46.6 cm) Height
# de catalogue 76502-608
Fournisseur: Eagle Manufacturing
Customers Who Bought This Also Bought
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Fournisseur: Saint Gobain Performance Plastics
1 747,88 $
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
1 098,06 $
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Fournisseur: VWR International
541,80 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
271,77 $
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
86,77 $ to 708,50 $