Lab Pack Open Head Poly Drum, 55 Gal, Metal Lever-Lock, 1x2in 1x3/4in Bung Holes, Yellow, Dimensions, Exterior23.75in (60.3 cm) Top, 19.38in (49.2 cm) Bottom, 39.125in (99.4 cm) Height
# de catalogue 76502-590
Fournisseur: Eagle Manufacturing
Customers Who Bought This Also Bought