VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Catalog # 46610-720A
Supplier: VWR International
Specifications
- Cap size:11 mm
- Septum:Red PTFE/White silicone
- Description:Snap caps with septa
- Material:PE
- Cat. no.:46610-720A
- Cap Color:Natural