ContactUsLinkComponent
VWR Snap Cap, Clear, 11 mm, Polyethylene, Clear PTFE/Synthetic Red Rubber
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
# de catalogue 46610-718A
Fournisseur:  VWR International
VWR Snap Cap, Clear, 11 mm, Polyethylene, Clear PTFE/Synthetic Red Rubber
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
# de catalogue 46610-718A
Fournisseur:  VWR International
Spécifications
  • Cap Size:
    11 mm
  • Septum:
    Clear PTFE/Synthetic red rubber
  • Description:
    Snap caps with septa
  • Material:
    PE
  • Cat. no.:
    46610-718A
  • Cap Color:
    Natural