ContactUsLinkComponent
VWR Cap, Open top Screw, Black, Polypropylene, Red PTFE/White Silicone, For 4ml Screw-Thread Vials
VWR® Screw Caps for 4 ml Screw-Thread Vials
Catalog # 46610-706A
Supplier:  VWR International
VWR Cap, Open top Screw, Black, Polypropylene, Red PTFE/White Silicone, For 4ml Screw-Thread Vials
VWR® Screw Caps for 4 ml Screw-Thread Vials
Catalog # 46610-706A
Supplier:  VWR International
Specifications
  • Cap size:
    13-425
  • Septum:
    Red PTFE/white silicone
  • Description:
    Screw caps with septa
  • Material:
    PP
  • Cat. no.:
    46610-706A
  • Cap Color:
    Black