VWR® Screw Caps for 8-425 Screw-Thread Vials, Standard Opening
Catalog # 46610-700A
Supplier: VWR International
Specifications
- Cap size:8-425
- Septum:Red PTFE/white silicone
- Description:Screw caps with septa
- Material:PP
- Cat. no.:46610-700A
- Cap Color:Black