VWR® Inserts for Wide Opening Vials
# de catalogue 82028-450A
Fournisseur: VWR International
Spécifications
- Cap included:No
- Capacity:0.33 ml
- Color:Clear
- Description:Insert with poly-support spring for wide opening vials
- Material:PP
- Cat. no.:82028-450A
- Bottom Style:Conical bottom