ContactUsLinkComponent
Pipet Tip, Conductive, Filter, Sterile, for Hamilton type systems, Size: 300Ul
J.T.Baker® Robotic Tips Type HT
# de catalogue 49008-1105
   Low Manufacturing Impact
J.T.Baker® Robotic Tips Type HT
# de catalogue 49008-1105
   Low Manufacturing Impact
Spécifications
  • Volume:
    300 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Tip Color:
    Black
  • Packed:
    12× 5 Racks, 96 each, blistered
  • Filter:
    +
  • Sterile:
    +
Spécifications
A propos de cet article
Suitable for use on third party robotic instruments Hamilton Microlab Star and Nimbus. Available in conductive version, with or without filter.
  • Manufactured under cleanroom conditions (cleanroom class 8)
  • State of the art quality control
  • Ideal for high-throughput analysis
  • Barcoded trays for automated identification
  • Packed in blister
J.T.Baker® robotic tips type HT are suitable for third party robotic instruments Hamilton Microlab Star, Nimbus, and other robotic instruments using Hamilton’s CO-RE pipetting head.
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Hamilton, Microlab Star™, Nimbus® and CO-RE® are registered trademarks of Hamilton company.
Customers Who Bought This Also Bought