Kit, CLIA Waived, Rapidly detect and differentiate between SARS-CoV-2, Flu A, and Flu B from one sample on one test device, Test Time: 15 min, Shelf Life: 16 month, No. Of Reactions: 150, Target Organism Class: Antigen
# de catalogue 76500-074
Fournisseur: BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.