T-spindle set, particularly suitable for the measurement of non-flowing samples, such as creams, pastes or gelatin, right accessory for electrica Helistand, Thread M2.5 LH, Sample volume: 500 ml, Sample container: 600mL beaker low, Dimensions (W x H x D): 160 x 82 x 270 mm
# de catalogue 76499-916
Fournisseur: IKA WORKS, INC. MX
Customers Who Bought This Also Bought