Centrifuge, With Timer And Speed Control, 6 X 10/15ml Tubes, Max Speed 4,000 rpm, 110V, 60Hz
# de catalogue 76499-386
Fournisseur: C & A Scientific
Centrifuge, With Timer And Speed Control, 6 X 10/15ml Tubes, Max Speed 4,000 rpm, 110V, 60Hz
# de catalogue 76499-386
Fournisseur: C & A Scientific
Customers Who Bought This Also Bought
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Fournisseur: VWR International
$89.92 to $1,797.48
VWR® Single Channel Electronic Timer with Memory and Clock and Certificate of Calibration
Fournisseur: VWR International
$75.03 to $739.18