ContactUsLinkComponent
Pipette Tip, Universal, With Filter, Low Retention, Sterile, Material: Polypropylene, polyethylene filter, which prevents liquids and aerosols from entering the body of pipette, feature a hydrophobic and smooth surface, Noncytotoxic and nonhemolytic, Size: 200Ul, 10RacksX96
Cole-Parmer® Universal Pipette Tips with Filter, Sterile
# de catalogue 76499-284
Fournisseur:  Argos Technologies
Pipette Tip, Universal, With Filter, Low Retention, Sterile, Material: Polypropylene, polyethylene filter, which prevents liquids and aerosols from entering the body of pipette, feature a hydrophobic and smooth surface, Noncytotoxic and nonhemolytic, Size: 200Ul, 10RacksX96
Cole-Parmer® Universal Pipette Tips with Filter, Sterile
# de catalogue 76499-284
Fournisseur:  Argos Technologies
Spécifications
  • Volume:
    200 µl
  • Tip Color:
    Transparent
  • Packed:
    10 Racks of 96 Tips
  • Filter:
    +
  • Sterile:
    +
Spécifications
A propos de cet article
Protect against cross-contamination with tips free of human touch.
  • Pure porous polyethylene filter protects against cross-contamination
  • Durable polypropylene construction
  • Noncytotoxic and nonhemolytic
  • Sterilized and DNase, RNase, and pyrogen free
Cole-Parmer sterile pipette tips are constructed of high-grade polypropylene and a polyethylene filter which prevents liquids and aerosols from entering the body of your pipette. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.
Packaging: Racks (96 tips per rack): These racks, with smooth rounded edges are extremely durable and a snap latch.
Customers Who Bought This Also Bought