Beaker, DWB 600, double-walled, has a filling volume of 600 ml. It can be connected to an external cooling or heating source and is therefore suitable for tempering the sample inside the beaker.
# de catalogue 76499-096
Fournisseur: IKA Works
Customers Who Bought This Also Bought